2-(Trifluoromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Trifluoromethyl)phenylboronic acid, (2-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethyl)phenyl)borane, 2-(Trifluoromethyl)benzeneboronic acid

BB-2625-5G BB-2625-25G
COBBBB-2625-5GEA 0 GBP
COBBBB-2625-5G COBBBB-2625-25G
2-(Trifluoromethyl)phenylboronic acid
2-(Trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
MDL Number: MFCD00236059
CAS Number: 1423-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR