VWR® Safetypette, Pipette Filler
Supplier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® Safetypette
612-4548EA 0 GBP
612-4548
VWR® Safetypette, Pipette Filler
Pipettes Pipette Controllers
Pipette filler for glass and plastic pipettes from 1 to 100 ml.
  • Replaceable hydrophobic membrane filter protects the instrument from liquid penetration
  • Separate buttons for aspirating/dispensing and discharging
  • Silicone adapter for leakproof positioning of each pipette
The Safetypette pipette filler facilitates the pipetting of a wide range of liquids. The practical arrangement of the controls makes work easier giving maximum safety during serial pipetting.
Ordering information: Supplied with three separate nose cones in three different colours.