Anaerobic systems, anaerobic jars
Supplier: Don Whitley Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Whitley
710-1213EA 0 GBP
710-1213 710-0841
Anaerobic systems, anaerobic jars
Anaerobic Systems
These anaerobic jars are robustly constructed in stainless steel and are simple to use.
  • Patented valve system
  • Fitted with the Oxoid low temperature catalyst sachet
They can be used with either gas generating envelopes or cylinder gas and will vent automatically if excess pressure is applied.
Ordering information: A large range of accessories are available. Please contact your local Avantor office for further information.
Delivery information: Supplied ready-to-use and complete with a comprehensive spares kit.