Norharmane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Norharman, 9H-Pyrido[3,4-b]indole

M-Clarity™ quality level = MQ200

N6252-100MG N6252-1G N6252-500MG
SIALN6252-100MGEA 86.6 GBP
SIALN6252-100MG SIALN6252-1G SIALN6252-500MG
Norharmane, Sigma-Aldrich®
Norharmane
Formula: C₁₁H₈N₂
MW: 168.2 g/mol
MDL Number: MFCD00004956
CAS Number: 244-63-3
EINECS: 205-959-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR