4-Quinazolone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinazolin-4(3H)-one4-Hydroxyquinazoline

M-Clarity™ quality level = MQ100

SIALH57807-5GEA 58.6 GBP
SIALH57807-5G
4-Quinazolone, Sigma-Aldrich®
4-Quinazolone
Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD00511302
CAS Number: 491-36-1
EINECS: 207-735-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR