4-Quinazolone 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Quinazolin-4(3H)-one

ACRO121880250EA 75.6 GBP
ACRO121880250
4-Quinazolone 98%
4-Quinazolone
Formula: C₈H₆N₂O
MW: 146.15 g/mol
Melting Pt: 215…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00511302
CAS Number: 491-36-1
EINECS: 207-735-8

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 215 to 219 °C
Titration with TBAH ≥97.5 %
Enantiomeric excess ≥97.5 %
Specific optical rotation -118° to -122° (20 °C, 589 nm) (c=5, in CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR