2-Ethyl-2-oxazoline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

137456-100ML 137456-500ML
SIAL137456-100MLEA 61 GBP
SIAL137456-100ML SIAL137456-500ML
2-Ethyl-2-oxazoline, Sigma-Aldrich®
2-Ethyl-2-oxazoline
Formula: C₅H₉NO
MW: 99.13 g/mol
MDL Number: MFCD00005302
CAS Number: 10431-98-8
EINECS: 233-912-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR