9-Fluorenylmethyl chloroformate (Fmoc chloride)

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 9H-Fluoren-9-ylmethyl chloroformate(9H-Fluoren-9-yl)methyl carbonochloridateFmoc-Cl9-Fluorenylmethyl chloroformateChloroformic acid 9-fluorenylmethyl ester, FMOCCl

MOLE72095944-5GEA 0 GBP
MOLE72095944-5G
9-Fluorenylmethyl chloroformate (Fmoc chloride)
9-Fluorenylmethyl chloroformate (Fmoc chloride)
Formula: C₁₅H₁₁ClO₂
MW: 258.7 g/mol
MDL Number: MFCD00001138
CAS Number: 28920-43-6
EINECS: 249-313-6
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR