1,2-Bis(phenylthio)ethane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003673-10G
003673-50G
FLUO003673-10GEA
0
GBP
FLUO003673-10G
FLUO003673-50G
1,2-Bis(phenylthio)ethane
1,2-Bis(phenylthio)ethane
Formula:
C₁₄H₁₄S₂ |
MDL Number:
MFCD00014085 CAS Number: 622-20-8 EINECS: 210-723-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...