Aluminon, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate)
, Aurin Tricarboxylic Acid Ammonium Salt
M-Clarity™ quality level = MQ100
1.00128.0010EA
741
GBP
1.00128.0010
1.00128.9999
Aluminon, Supelco®
Aluminon
Formula:
C₂₂H₂₃N₃O₉ MW: 473.44 g/mol |
MDL Number:
MFCD00040925 CAS Number: 569-58-4 EINECS: 209-319-1 |
Specification Test Results
Solubility | 800 g/l (20 °C) |
Melting Point | 220 - 225 °C decomposes |
Molar Mass | 473.44 g/mol |
Bulk Density | 400 kg/m3 |
pH | 5.3 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...