3-Amino-1H-pyrazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole

M-Clarity™ quality level = MQ100

160644-10G 160644-2.5G
SIAL160644-10GEA 353 GBP
SIAL160644-10G SIAL160644-2.5G
3-Amino-1H-pyrazole, Sigma-Aldrich®
3-Amino-1H-pyrazole
Formula: C₃H₅N₃
MW: 83.09 g/mol
MDL Number: MFCD00005236
CAS Number: 1820-80-0
EINECS: 217-338-1
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR