Tetrafluorohydroquinone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3,5,6-Tetrafluorohydroquinone, Tetrafluorobenzene-1,4-diol

007363-25G 007363-5G
FLUO007363-25GEA 0 GBP
FLUO007363-25G FLUO007363-5G
Tetrafluorohydroquinone
Tetrafluorohydroquinone
Formula: C₆H₂F₄O₂
MW: 182.07 g/mol
MDL Number: MFCD00002340
CAS Number: 771-63-1
EINECS: 212-237-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR