L(+)-Ornithine hydrochloride

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Orn-OH·HClH-Orn-OH Hydrochloride

MOLE11702720-1KGEA 0 GBP
MOLE11702720-1KG
L(+)-Ornithine hydrochloride
L(+)-Ornithine hydrochloride
Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
MDL Number: MFCD00064562
CAS Number: 3184-13-2
EINECS: 221-678-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR