Tungsten hexacarbonyl ≥99% (<0.3%-Mo)

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Tungsten carbonylHexacarbonyltungsten

STRE74-2200-25GEA 0 GBP
STRE74-2200-25G
Tungsten hexacarbonyl ≥99% (<0.3%-Mo)
Tungsten hexacarbonyl

Metal Carbonyls, Volatile Metal Carbonyls for CVD & ALD, Volatile Precursors for CVD

Formula: C₆O₆W
MW: 351.9 g/mol
Boiling Pt: 451.3 °C (1013 hPa)
Melting Pt: 169…170 °C (dec.)
Density: 2.65 g/cm³ (20 °C)
MDL Number: MFCD00011462
CAS Number: 14040-11-0
EINECS: 237-880-2
UN: 3466
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR