Phenyltrimethoxysilane

Supplier: ABCR GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Trimethoxy(phenyl)silane

ABCRAB111342.0100EA 0 GBP
ABCRAB111342.0100
Phenyltrimethoxysilane
Phenyltrimethoxysilane
Formula: C₉H₁₄O₃Si
MW: 198.29 g/mol
Density: 1.064 g/cm³ (20 °C)
MDL Number: MFCD00025689
CAS Number: 2996-92-1
EINECS: 221-066-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR