Methyltrichlorosilane

Supplier: ABCR GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Trichloro(methyl)silane

ABCRAB129194.0500EA 0 GBP
ABCRAB129194.0500 ABCRAB129194.0025
Methyltrichlorosilane
Methyltrichlorosilane
Formula: CH₃Cl₃Si
MW: 149.48 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –90 °C
Density: 1.27 g/cm³ (20 °C)
Flash Pt: 1 °C
MDL Number: MFCD00000481
CAS Number: 75-79-6
EINECS: 200-902-6
UN: 1250
ADR: 3,I
REACH: 01-2119475226-36

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR