5-Bromo-2-mercaptobenzothiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1554-1G HI-1554-5G
COBBHI-1554-1GEA 0 GBP
COBBHI-1554-1G COBBHI-1554-5G
5-Bromo-2-mercaptobenzothiazole
5-Bromo-2-mercaptobenzothiazole
Formula: C₇H₄BrNS₂
MDL Number: MFCD08460052
CAS Number: 71216-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR