4-Bromo-2-mercaptobenzothiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1561-1G HI-1561-5G
COBBHI-1561-1GEA 0 GBP
COBBHI-1561-1G COBBHI-1561-5G
4-Bromo-2-mercaptobenzothiazole
4-Bromo-2-mercaptobenzothiazole
Formula: C₇H₄BrNS₂
MDL Number: MFCD11044824
CAS Number: 1083181-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR