4-Bromo-2-mercapto-6-(trifluoromethoxy)benzothiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1242-5G PY-1242-1G
COBBPY-1242-5GEA 0 GBP
COBBPY-1242-5G COBBPY-1242-1G
4-Bromo-2-mercapto-6-(trifluoromethoxy)benzothiazole
4-Bromo-2-mercapto-6-(trifluoromethoxy)benzothiazole
Formula: C₈H₃BrF₃NOS₂
MDL Number: MFCD13195780
CAS Number: 1215206-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR