4-Bromo-6-chloro-2-mercaptobenzothiazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-3136-5G
HC-3136-1G
COBBHC-3136-5GEA
0
GBP
COBBHC-3136-5G
COBBHC-3136-1G
4-Bromo-6-chloro-2-mercaptobenzothiazole
4-Bromo-6-chloro-2-mercaptobenzothiazole
Formula:
C₇H₃BrClNS₂ |
MDL Number:
MFCD15143552 CAS Number: 1215205-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...