Tetraphenylphosphonium hexafluoroantimonate ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B24758.06 B24758.14
B24758.06EA 0 GBP
B24758.06 B24758.14
Tetraphenylphosphonium hexafluoroantimonate ≥98%
Tetraphenylphosphonium hexafluoroantimonate

Purity: 98+% CAS No.: 124329-50-6 Molecular Formula: (C5H5)4PSbF6 Formula Weight: 575.13 MDL No.: MFCD00051888

Formula: C₂₄H₂₀F₆PSb
MW: 575.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051888
CAS Number: 124329-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR