3-Cyclopropylpropionyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Cyclopropylpropanoic acid
011665-5G
011665-1G
FLUO011665-5GEA
0
GBP
FLUO011665-5G
FLUO011665-1G
3-Cyclopropylpropionyl chloride
3-Cyclopropylpropionic acid
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol |
MDL Number:
MFCD02179408 CAS Number: 5618-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...