3-Chlorothiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001747-25G
001747-100G
FLUO001747-25GEA
0
GBP
FLUO001747-25G
FLUO001747-100G
3-Chlorothiophene
3-Chlorothiophene
Formula:
C₄H₃ClS MW: 118.59 g/mol Boiling Pt: 133…135 °C (1013 hPa) Density: 1.276 g/cm³ (20 °C) |
MDL Number:
MFCD00043887 CAS Number: 17249-80-8 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...