2-Chlorothiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Thienyl chloride
002954-100G
002954-500G
FLUO002954-100GEA
0
GBP
FLUO002954-100G
FLUO002954-500G
2-Chlorothiophene
2-Chlorothiophene
Formula:
C₄H₃ClS MW: 118.59 g/mol Boiling Pt: 127 °C (1013 hPa) Melting Pt: –1 °C Density: 1.285 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005421 CAS Number: 96-43-5 EINECS: 202-505-3 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...