2-Bromo-5-chlorothiophene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B2620.5 B2620.25
XXXXB2620.5EA 0 GBP
XXXXB2620.5 XXXXB2620.25
2-Bromo-5-chlorothiophene
2-Bromo-5-chlorothiophene
Formula: C₄H₂BrClS
MW: 197.48 g/mol
Boiling Pt: 45…46 °C (1 mmHg)
Melting Pt: –2 °C
Density: 1.803 g/cm³ (25 °C)
MDL Number: MFCD00014523
CAS Number: 2873-18-9
EINECS: 220-708-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR