2-Bromo-5-chlorothiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001106-25G
001106-100G
001106-5G
FLUO001106-25GEA
0
GBP
FLUO001106-25G
FLUO001106-100G
FLUO001106-5G
2-Bromo-5-chlorothiophene
2-Bromo-5-chlorothiophene
Formula:
C₄H₂BrClS MW: 197.48 g/mol Boiling Pt: 45…46 °C (1 mmHg) Melting Pt: –2 °C Density: 1.803 g/cm³ (25 °C) |
MDL Number:
MFCD00014523 CAS Number: 2873-18-9 EINECS: 220-708-5 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...