2,3,5,6-Tetrafluoropyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
009346-1G
009346-5G
009346-10G
FLUO009346-1GEA
0
GBP
FLUO009346-1G
FLUO009346-5G
FLUO009346-10G
2,3,5,6-Tetrafluoropyridine
2,3,5,6-Tetrafluoropyridine
Formula:
C₅HF₄N MW: 151.06 g/mol Boiling Pt: 98…99 °C (1013 hPa) Density: 1.499 g/cm³ (20 °C) |
MDL Number:
MFCD00011734 CAS Number: 2875-18-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...