Tetrahydrofuran
Supplier: MARKET SOURCE PART PROCESS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Butylene oxide
, THF
, Diethylene oxide
XXXXRH1011-2.5EA
73.2
GBP
XXXXRH1011-2.5
IBSARH1011
MSPPRH1011GSK-1
Tetrahydrofuran
Tetrahydrofuran
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 66 °C (1013 hPa) Melting Pt: –108.5 °C Density: 0.888 g/cm³ (20 °C) Flash Pt: –21.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005356 CAS Number: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3,II REACH: 05-2114615188-48 Merck Index: 12,09351 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...