1,4-Dimethylnaphthalene
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC20760000EA
60.1
GBP
EHERC20760000
1,4-Dimethylnaphthalene
1,4-Dimethylnaphthalene
Formula:
C₁₂H₁₂ MW: 156.23 g/mol Boiling Pt: 264 °C (1013 hPa) Density: 1.016 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004037 CAS Number: 571-58-4 EINECS: 209-335-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...