Decacarbonyldirhenium ≥96%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Rhenium carbonyl
62108.03EA
110
GBP
62108.03
Decacarbonyldirhenium ≥96%
Decacarbonyldirhenium
Formula:
C₁₀O₁₀Re₂ MW: 652.52 g/mol Melting Pt: 170 °C (dec.) Density: 2.87 Storage Temperature: Ambient |
MDL Number:
MFCD00011198 CAS Number: 14285-68-8 EINECS: 238-202-8 UN: 3466 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...