Decacarbonyldirhenium, sublimated

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Rhenium carbonyl

ACRO223990050EA 458 GBP
ACRO223990050
Decacarbonyldirhenium, sublimated
Decacarbonyldirhenium
Formula: C₁₀O₁₀Re₂
MW: 652.52 g/mol
Melting Pt: 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011198
CAS Number: 14285-68-8
EINECS: 238-202-8
UN: 3466
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow-green Crystals
Infrared spectrum Conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR