5-Bromoisoquinoline

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

MOLE48905952-1GEA 0 GBP
MOLE48905952-1G
5-Bromoisoquinoline
5-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
MDL Number: MFCD01646405
CAS Number: 34784-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR