Phenylacetamide

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Phenylacetamide

P1325.25 P1325.100
XXXXP1325.25EA 0 GBP
XXXXP1325.25 XXXXP1325.100
Phenylacetamide
Phenylacetamide
Formula: C₈H₉NO
MW: 135.17 g/mol
MDL Number: MFCD00059193
CAS Number: 103-81-1
EINECS: 203-147-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR