5-Nitroisatoic anhydride

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione

N1527.5 N1527.25
XXXXN1527.5EA 0 GBP
XXXXN1527.5 XXXXN1527.25
5-Nitroisatoic anhydride
5-Nitroisatoic anhydride
Formula: C₈H₄N₂O₅
MW: 208.13 g/mol
MDL Number: MFCD00016919
CAS Number: 20829-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR