2,5-Dichloroisonicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,5-Dichloropyridine-4-carboxylic acid

D2599-500MG D2599.500MG D2599-5G
TCIAD2599-500MGEA 0 GBP
TCIAD2599-500MG FLUOD2599.500MG TCIAD2599-5G
2,5-Dichloroisonicotinic acid ≥98.0% (by GC, titration analysis)
2,5-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00466640
CAS Number: 88912-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2fc6aa75d052ac516dd5c304ce3f5bac27b7d4e&vid=f2fc6aa75d052ac516dd5c304ce3f5bac27b7d4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR