2-Bromoethyl ethyl ether, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B66205-100G B66205-25G
SIALB66205-100GEA 183 GBP
SIALB66205-100G SIALB66205-25G
2-Bromoethyl ethyl ether, Sigma-Aldrich®
2-Bromoethyl ethyl ether
Formula: C₄H₉BrO
MW: 153.02 g/mol
MDL Number: MFCD00000237
CAS Number: 592-55-2
EINECS: 209-763-6
UN: 2340
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR