Bromoferrocene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SIAL725242-5GEA 282 GBP
SIAL725242-5G SIAL725242-2G
Bromoferrocene, Sigma-Aldrich®
Bromoferrocene
Formula: C₁₀H₉BrFe
MDL Number: MFCD03093913
CAS Number: 1273-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR