Methyl-2-aminonicotinate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 2-aminonicotinate
715107-1G
715107-5G
SIAL715107-1GEA
39.3
GBP
SIAL715107-1G
SIAL715107-5G
Methyl-2-aminonicotinate, Sigma-Aldrich®
Methyl 2-aminonicotinate
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol |
MDL Number:
MFCD00128872 CAS Number: 14667-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...