Methyl-6-aminonicotinate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester

648736-1G 648736-5G
SIAL648736-1GEA 70.3 GBP
SIAL648736-1G SIAL648736-5G
Methyl-6-aminonicotinate, Sigma-Aldrich®
Methyl 6-aminonicotinate
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD00797844
CAS Number: 36052-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR