Lead acetate cotton Reag. Ph. Eur. 1048101
Supplier: REAGECON
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
Lead di-acetate
, Lead (II) acetate on cotton
REGE1048101EA
311
GBP
REGE1048101
Lead acetate cotton Reag. Ph. Eur. 1048101
Lead acetate cotton
- Composition: 1 dilute acetic acid:10 lead acetate solution
- Form: Cottony balls
Manufactured according to European Pharmacopoeia Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Lead Acetate Cotton, EP Grade, Reagecon™ is a ready-to-use, dried cotton that has absorbed a 10:1 lead acetate-dilute acetic acid solution for use in arsenic limit tests.
Formula:
(H₃CCOO)₂Pb MW: 325.29 g/mol |
CAS Number:
6080-56-4 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...