5-Aminopicolinic acid

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid

87232213/1KG 87232213/500G 87232213/2KG
MOLE87232213/1KGEA 0 GBP
MOLE87232213/1KG MOLE87232213/500G MOLE87232213/2KG
5-Aminopicolinic acid
5-Aminopicolinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
MDL Number: MFCD02684600
CAS Number: 24242-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR