4-Nitroisatoic anhydride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 7-Nitro-1H-benzo[d][1,3]oxazine-2,4-dione

011830-10G 011830-5G
FLUO011830-10GEA 0 GBP
FLUO011830-10G FLUO011830-5G
4-Nitroisatoic anhydride
4-Nitroisatoic anhydride
Formula: C₈H₄N₂O₅
MW: 208.13 g/mol
MDL Number: MFCD00798220
CAS Number: 63480-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR