N-Butylbenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Benzyl-n-butylamine

001047-25G 001047-100G
FLUO001047-25GEA 0 GBP
FLUO001047-25G FLUO001047-100G
N-Butylbenzylamine
N-Butylbenzylamine
Formula: C₁₁H₁₇N
MDL Number: MFCD00009427
CAS Number: 2403-22-7
EINECS: 219-287-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR