Methyl-6-iodonicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 6-iodonicotinate

PY-8010-1G PY-8010-5G
COBBPY-8010-1GEA 0 GBP
COBBPY-8010-1G COBBPY-8010-5G
Methyl-6-iodonicotinate
Methyl 6-iodonicotinate
Formula: C₇H₆INO₂
MW: 263.03 g/mol
MDL Number: MFCD12546492
CAS Number: 173157-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR