Methyl-5-bromo-2-chloronicotinate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 5-bromo-2-chloronicotinate
PY-7076-1G
PY-7076-5G
COBBPY-7076-1GEA
0
GBP
COBBPY-7076-1G
COBBPY-7076-5G
Methyl-5-bromo-2-chloronicotinate
Methyl 5-bromo-2-chloronicotinate
Formula:
C₇H₅BrClNO₂ MW: 250.48 g/mol |
MDL Number:
MFCD03844848 CAS Number: 78686-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...