6-Chloropicolinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Chloropyridine-2-carboxylic acid, 2-Chloropyridine-6-carboxylic acid

CA-4246-5G CA-4246-25G
COBBCA-4246-5GEA 0 GBP
COBBCA-4246-5G COBBCA-4246-25G
6-Chloropicolinic acid
6-Chloropicolinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD00155390
CAS Number: 4684-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR