Methyl-5-bromo-2-methoxynicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 5-bromo-2-methoxynicotinate, Methyl 5-bromo-2-methoxy-3-pyridinecarboxylate

CA-4186-5G CA-4186-1G
COBBCA-4186-5GEA 0 GBP
COBBCA-4186-5G COBBCA-4186-1G
Methyl-5-bromo-2-methoxynicotinate
Methyl 5-bromo-2-methoxynicotinate
Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
MDL Number: MFCD08064227
CAS Number: 122433-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR