2,5-Dichloroisonicotinic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Dichloropyridine-4-carboxylic acid

3W-0027.500 3W-0027.1
BIOT3W-0027.500EA 0 GBP
BIOT3W-0027.500 BIOT3W-0027.1
2,5-Dichloroisonicotinic acid
2,5-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00466640
CAS Number: 88912-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR