5-Bromopyridine-2-carbonitrile 95%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Bromopicolinonitrile, 5-Bromo-2-cyanopyridine

L19621.06 L19621.03
L19621.06EA 40.8 GBP
L19621.06 L19621.03
5-Bromopyridine-2-carbonitrile 95%
5-Bromopyridine-2-carbonitrile

Purity: 95% CAS No.: 97483-77-7 Molecular Formula: C6H3BrN2 Formula Weight: 183.01 Melting Point: 124-127° Beilstein Reference: 5498624

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Melting Pt: 124…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00234144
CAS Number: 97483-77-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR