4-Chloro-3-iodobenzoic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C2211-D1.5 C2211-D1.25
XXXXC2211-D1.5EA 0 GBP
XXXXC2211-D1.5 XXXXC2211-D1.25
4-Chloro-3-iodobenzoic acid
4-Chloro-3-iodobenzoic acid
Formula: C₇H₄ClIO₂
MDL Number: MFCD00079732
CAS Number: 42860-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR